Assay Details
| Gene Symbol: RPL7L1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005627364.3 | XP_005627421.1 | 5 - 6 | 813 | 65 | |
| XM_538924.6 | XP_538924.3 | 5 - 6 | 824 | 65 | ||
More Information
Set Membership: |
|

RPL7L1
Genomic Map