Assay Details
| Gene Symbol: PPP2R1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005616222.3 | XP_005616279.1 | 8 - 9 | 1100 | 107 | |
| XM_845900.5 | XP_850993.1 | 8 - 9 | 1100 | 107 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

PPP2R1A
Genomic Map