Assay Details
| Gene Symbol: POLR3F | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005634741.3 | XP_005634798.1 | 7 - 8 | 730 | 120 | |
| XM_005634742.3 | XP_005634799.1 | 7 - 8 | 686 | 120 | ||
| XM_852045.5 | XP_857138.2 | 6 - 7 | 709 | 120 | ||
More Information
Set Membership: |
|

POLR3F
Genomic Map