Assay Details
| Gene Symbol: SOCS3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001031631.1 | NP_001026801.1 | 164 | 128 | ||
| GenBank mRNA | AB164434.1 | - | 164 | 128 | ||
| AY485997.1 | - | 116 | 128 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

SOCS3
Genomic Map