Assay Details
| Gene Symbol: Ccm2l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007642935.1 | XP_007641125.1 | 1 - 2 | 29 | 66 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ccm2l| Gene Symbol: Ccm2l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007642935.1 | XP_007641125.1 | 1 - 2 | 29 | 66 | |
Set Membership: |
|