Assay Details
| Gene Symbol: LOC100769029 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003510677.2 | XP_003510725.1 | 1 - 2 | 60 | 74 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100769029| Gene Symbol: LOC100769029 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003510677.2 | XP_003510725.1 | 1 - 2 | 60 | 74 | |