Assay Details
| Gene Symbol: Ndufb8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007649075.1 | XP_007647265.1 | 2 - 3 | 159 | 100 | |
| GenBank mRNA | JI882653.1 | - | 1 - 2 | 104 | 100 | |
More Information
Set Membership: |
|

Ndufb8
Genomic Map