Assay Details
| Gene Symbol: Mrpl10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003499569.2 | XP_003499617.1 | 2 - 3 | 257 | 84 | |
| GenBank mRNA | JI896193.1 | - | 2 - 3 | 276 | 84 | |
| JP054609.1 | - | 2 - 3 | 276 | 84 | ||
More Information
Set Membership: |
|

Mrpl10
Genomic Map