Assay Details
| Gene Symbol: Rpl7l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003506052.2 | XP_003506100.1 | 6 - 7 | 1043 | 59 | |
| GenBank mRNA | JI862859.1 | - | 6 - 7 | 788 | 59 | |
| JP050020.1 | - | 6 - 7 | 788 | 59 | ||
More Information
Set Membership: |
|

Rpl7l1
Genomic Map