Assay Details
| Gene Symbol: Clcf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003509939.2 | XP_003509987.1 | 2 - 3 | 405 | 84 | |
| XM_007650597.1 | XP_007648787.1 | 2 - 3 | 505 | 84 | ||
| XM_007650598.1 | XP_007648788.1 | 2 - 3 | 508 | 84 | ||
| GenBank mRNA | JI882866.1 | - | 1 - 2 | 165 | 84 | |
| JP042429.1 | - | 1 - 2 | 153 | 84 | ||
More Information
Set Membership: |
|

Clcf1
Genomic Map