Assay Details
| Gene Symbol: NDUFB8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005698396.3 | XP_005698453.2 | 3 - 4 | 378 | 70 | |
| XM_018041257.1 | XP_017896746.1 | 3 - 4 | 363 | 70 | ||
| GenBank mRNA | JO517302.2 | - | 2 - 3 | 344 | 70 | |
| JO537207.2 | - | 2 - 3 | 343 | 70 | ||
More Information
Set Membership: |
|

NDUFB8
Genomic Map