Assay Details
| Gene Symbol: PDGFB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005681085.3 | XP_005681142.3 | 4 - 5 | 499 | 87 | |
| XM_013964353.2 | XP_013819807.2 | 4 - 5 | 499 | 87 | ||
| XM_018048784.1 | XP_017904273.1 | 4 - 5 | 499 | 87 | ||
More Information
Set Membership: |
|

PDGFB
Genomic Map