Assay Details
| Gene Symbol: MILR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005694010.3 | XP_005694067.2 | 1 - 2 | 275 | 73 | |
| XM_013972411.2 | XP_013827865.2 | 1 - 2 | 253 | 73 | ||
| XM_018065359.1 | XP_017920848.1 | 1 - 2 | 275 | 73 | ||
| GenBank mRNA | JO595577.2 | - | 1 - 2 | 185 | 73 | |
| JO595578.2 | - | 1 - 2 | 185 | 73 | ||
More Information
Set Membership: |
|

MILR1
Genomic Map