Assay Details
| Gene Symbol: BCAR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018061715.1 | XP_017917204.1 | 1 - 2 | 2543 | 88 | |
| XM_018061717.1 | XP_017917206.1 | 2 - 3 | 702 | 88 | ||
| XM_018061718.1 | XP_017917207.1 | 2 - 3 | 735 | 88 | ||
| XM_018061719.1 | XP_017917208.1 | 2 - 3 | 630 | 88 | ||
| GenBank mRNA | JO637950.2 | - | 2 - 3 | 672 | 88 | |
More Information
Set Membership: |
|

BCAR1
Genomic Map