Assay Details
| Gene Symbol: PRSS8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018040628.1 | XP_017896117.1 | 3 - 4 | 527 | 58 | |
| XM_018040629.1 | XP_017896118.1 | 3 - 4 | 524 | 58 | ||
More Information
Set Membership: |
|

PRSS8
Genomic Map