Assay Details
| Gene Symbol: PRSS27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018040362.1 | XP_017895851.1 | 4 - 5 | 611 | 99 | |
| GenBank mRNA | KA339530.1 | - | 4 - 5 | 681 | 99 | |
More Information
Set Membership: |
|

PRSS27
Genomic Map