Assay Details
| Gene Symbol: RIMKLB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018048508.1 | XP_017903997.1 | 5 - 6 | 876 | 65 | |
| XM_018048509.1 | XP_017903998.1 | 5 - 6 | 876 | 65 | ||
| XM_018048510.1 | XP_017903999.1 | 5 - 6 | 876 | 65 | ||
| XM_018048511.1 | XP_017904000.1 | 5 - 6 | 876 | 65 | ||
| XM_018048512.1 | XP_017904001.1 | 5 - 6 | 876 | 65 | ||
| XM_018048513.1 | XP_017904002.1 | 5 - 6 | 1036 | 65 | ||
| XM_018048514.1 | XP_017904003.1 | 5 - 6 | 988 | 65 | ||
| XM_018048515.1 | XP_017904004.1 | 5 - 6 | 862 | 65 | ||
| XM_018048516.1 | XP_017904005.1 | 5 - 6 | 834 | 65 | ||
| XM_018048517.1 | XP_017904006.1 | 5 - 6 | 841 | 65 | ||
| XM_018048518.1 | XP_017904007.1 | 5 - 6 | 794 | 65 | ||
More Information
Set Membership: |
|

RIMKLB
Genomic Map