Assay Details
Gene Symbol: TREX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008981595.2 | XP_008979843.1 | 1 - 2 | 94 | 82 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_008981595.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
