Assay Details
| Gene Symbol: MRPL10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008997455.2 | XP_008995703.1 | 2 - 3 | 281 | 67 | |
| XM_008997456.2 | XP_008995704.1 | 2 - 3 | 403 | 67 | ||
More Information
Set Membership: |
|

MRPL10
Genomic Map