Assay Details
Gene Symbol: CPLANE2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002750350.3 | XP_002750396.2 | 2 - 3 | 265 | 106 | |
XM_017974358.1 | XP_017829847.1 | 2 - 3 | 302 | 106 |

More Information
Set Membership: |
![]() ![]() |
