Assay Details
| Gene Symbol: POLR3F | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002747475.4 | XP_002747521.1 | 4 - 5 | 431 | 106 | |
| XM_008995806.2 | XP_008994054.1 | 5 - 6 | 454 | 106 | ||
| XM_008995807.2 | XP_008994055.1 | 5 - 6 | 311 | 106 | ||
| XM_008995809.2 | XP_008994057.1 | 5 - 6 | 457 | 106 | ||
| XM_017971982.1 | XP_017827471.1 | 3 - 4 | 1046 | 106 | ||
More Information
Set Membership: |
|

POLR3F
Genomic Map