Assay Details
| Gene Symbol: Dscam4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001043131.3 | NP_001036596.2 | 28 - 29 | 5522 | 56 | |
| NM_001144436.2 | NP_001137908.1 | 28 - 29 | 5806 | 56 | ||
| NM_001144437.2 | NP_001137909.1 | 28 - 29 | 5806 | 56 | ||
| NM_001274642.1 | NP_001261571.1 | 28 - 29 | 5788 | 56 | ||
| NM_001274643.2 | NP_001261572.1 | 28 - 29 | 5678 | 56 | ||
More Information
Set Membership: |
|

Dscam4
Genomic Map