Assay Details
| Gene Symbol: Dscam1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001043033.1 | NP_001036498.1 | 4 - 5 | 1090 | 82 | |
| NM_001043044.1 | NP_001036509.1 | 4 - 5 | 1090 | 82 | ||
| NM_001043055.1 | NP_001036520.1 | 4 - 5 | 1090 | 82 | ||
| NM_001259233.1 | NP_001246162.1 | 4 - 5 | 1090 | 82 | ||
| NM_001259234.1 | NP_001246163.1 | 4 - 5 | 1090 | 82 | ||
| NM_001259235.1 | NP_001246164.1 | 4 - 5 | 1090 | 82 | ||
| NM_001259239.1 | NP_001246168.1 | 4 - 5 | 1090 | 82 | ||
| NM_001259247.1 | NP_001246176.1 | 4 - 5 | 1090 | 82 | ||
| NM_001259249.1 | NP_001246178.1 | 4 - 5 | 1090 | 82 | ||
| NM_001259252.1 | NP_001246181.1 | 4 - 5 | 1090 | 82 | ||
| NM_001273835.1 | NP_001260764.1 | 4 - 5 | 1120 | 82 | ||
| NM_165513.1 | NP_724542.1 | 4 - 5 | 1090 | 82 | ||
| NM_165515.2 | NP_724544.2 | 4 - 5 | 1090 | 82 | ||
More Information
Set Membership: |
|

Dscam1
Genomic Map