Assay Details
| Gene Symbol: Dscam1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_078925.5 | NP_523649.5 | 5 - 6 | 1250 | 63 | |
| NM_165513.1 | NP_724542.1 | 5 - 6 | 1241 | 63 | ||
| NM_165514.1 | NP_724543.1 | 5 - 6 | 1241 | 63 | ||
More Information
Set Membership: |
|

Dscam1
Genomic Map