Assay Details
| Gene Symbol: Dscam1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001043020.1 | NP_001036485.1 | 9 - 10 | 2668 | 68 | |
| NM_001043057.1 | NP_001036522.1 | 9 - 10 | 2659 | 68 | ||
| NM_078925.5 | NP_523649.5 | 9 - 10 | 2665 | 68 | ||
| NM_165513.1 | NP_724542.1 | 9 - 10 | 2656 | 68 | ||
| NM_165514.1 | NP_724543.1 | 9 - 10 | 2656 | 68 | ||
More Information
Set Membership: |
|

Dscam1
Genomic Map