Assay Details
| Gene Symbol: Dscam1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001043022.1 | NP_001036487.1 | 4 - 5 | 1087 | 71 | |
| NM_001043029.1 | NP_001036494.1 | 4 - 5 | 1087 | 71 | ||
| NM_001043047.1 | NP_001036512.1 | 4 - 5 | 1087 | 71 | ||
| NM_001043049.1 | NP_001036514.1 | 4 - 5 | 1087 | 71 | ||
| NM_001043058.1 | NP_001036523.1 | 4 - 5 | 1087 | 71 | ||
| NM_001043060.1 | NP_001036525.1 | 4 - 5 | 1087 | 71 | ||
| NM_165514.1 | NP_724543.1 | 4 - 5 | 1087 | 71 | ||
More Information
Set Membership: |
|

Dscam1
Genomic Map