Assay Details
| Gene Symbol: Cngl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001258761.3 | NP_001245690.1 | 13 - 14 | 4664 | 64 | |
| NM_001258762.3 | NP_001245691.1 | 14 - 15 | 4709 | 64 | ||
More Information
Set Membership: |
|

Cngl
Genomic Map