Assay Details
| Gene Symbol: Cngl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001258761.3 | NP_001245690.1 | 15 - 16 | 6266 | 93 | |
| NM_001258762.3 | NP_001245691.1 | 16 - 17 | 6311 | 93 | ||
| NM_001272642.2 | NP_001259571.2 | 16 - 17 | 5700 | 93 | ||
| NM_001370020.1 | NP_001356908.1 | 16 - 17 | 5700 | 93 | ||
| NM_167441.5 | NP_727848.3 | 17 - 18 | 6311 | 93 | ||
More Information
Set Membership: |
|

Cngl
Genomic Map