Assay Details
| Gene Symbol: Cngl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001258762.3 | NP_001245691.1 | 4 - 5 | 2716 | 64 | |
| NM_001272642.2 | NP_001259571.2 | 4 - 5 | 2189 | 64 | ||
| NM_001370020.1 | NP_001356908.1 | 4 - 5 | 2189 | 64 | ||
| NM_167441.5 | NP_727848.3 | 4 - 5 | 2716 | 64 | ||
More Information
Set Membership: |
|

Cngl
Genomic Map