Assay Details
| Gene Symbol: Cngl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001258761.3 | NP_001245690.1 | 9 - 10 | 3544 | 119 | |
| NM_001258762.3 | NP_001245691.1 | 10 - 11 | 3589 | 119 | ||
| NM_001272642.2 | NP_001259571.2 | 10 - 11 | 3062 | 119 | ||
| NM_001370020.1 | NP_001356908.1 | 10 - 11 | 3062 | 119 | ||
More Information
Set Membership: |
|

Cngl
Genomic Map