Assay Details
| Gene Symbol: Cngl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001258761.3 | NP_001245690.1 | 10 - 11 | 3705 | 76 | |
| NM_001258762.3 | NP_001245691.1 | 11 - 12 | 3750 | 76 | ||
| NM_001272642.2 | NP_001259571.2 | 11 - 12 | 3223 | 76 | ||
| NM_001370020.1 | NP_001356908.1 | 11 - 12 | 3223 | 76 | ||
| NM_167441.5 | NP_727848.3 | 12 - 13 | 3834 | 76 | ||
More Information
Set Membership: |
|

Cngl
Genomic Map