Assay Details
| Gene Symbol: Dscam1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001043019.1 | NP_001036484.1 | 3 - 4 | 921 | 76 | |
| NM_001043021.1 | NP_001036486.1 | 3 - 4 | 921 | 76 | ||
| NM_001043023.1 | NP_001036488.1 | 3 - 4 | 921 | 76 | ||
| NM_001043027.1 | NP_001036492.1 | 3 - 4 | 921 | 76 | ||
| NM_001043051.1 | NP_001036516.1 | 3 - 4 | 921 | 76 | ||
| NM_001259245.1 | NP_001246174.1 | 3 - 4 | 921 | 76 | ||
| NM_001259250.1 | NP_001246179.1 | 3 - 4 | 921 | 76 | ||
| NM_078925.5 | NP_523649.5 | 3 - 4 | 921 | 76 | ||
More Information
Set Membership: |
|

Dscam1
Genomic Map