Assay Details
| Gene Symbol: babo | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_057651.4 | NP_476999.1 | 4 - 5 | 1331 | 65 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

babo| Gene Symbol: babo | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_057651.4 | NP_476999.1 | 4 - 5 | 1331 | 65 | |
Set Membership: |
|