Assay Details
| Gene Symbol: babo | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_057652.4 | NP_477000.1 | 4 - 5 | 1263 | 135 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

babo| Gene Symbol: babo | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_057652.4 | NP_477000.1 | 4 - 5 | 1263 | 135 | |
Set Membership: |
|