Assay Details
| Gene Symbol: Nrk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_057907.4 | NP_477255.1 | 3 - 4 | 1120 | 68 | |
| GenBank mRNA | BT033065.1 | - | 3 - 4 | 1107 | 68 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nrk| Gene Symbol: Nrk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_057907.4 | NP_477255.1 | 3 - 4 | 1120 | 68 | |
| GenBank mRNA | BT033065.1 | - | 3 - 4 | 1107 | 68 | |
Set Membership: |
|