Assay Details
| Gene Symbol: 140up | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001275632.1 | NP_001262561.1 | 1 - 2 | 249 | 74 | |
| NM_057603.6 | NP_476951.1 | 1 - 2 | 249 | 74 | ||
| GenBank mRNA | AY058577.1 | - | 1 - 2 | 189 | 74 | |
More Information
Set Membership: |
|

140up
Genomic Map