Assay Details
| Gene Symbol: mRpL10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_078716.3 | NP_523440.1 | 2 | 738 | 55 | |
| GenBank mRNA | AY060877.1 | - | 2 | 701 | 55 | |
| AY113438.1 | - | 2 | 733 | 55 | ||
More Information
Set Membership: |
|

mRpL10
Genomic Map