Assay Details
| Gene Symbol: crfb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077626.2 | NP_001071094.2 | 3 - 4 | 569 | 111 | |
| XM_005167960.4 | XP_005168017.1 | 3 - 4 | 664 | 111 | ||
| GenBank mRNA | EF014953.1 | - | 3 - 4 | 569 | 111 | |
More Information
Set Membership: |
|

crfb2
Genomic Map