Assay Details
| Gene Symbol: crfb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077626.2 | NP_001071094.2 | 4 - 5 | 722 | 90 | |
| GenBank mRNA | EF014953.1 | - | 4 - 5 | 722 | 90 | |
More Information
Set Membership: |
|

crfb2
Genomic Map