Assay Details
| Gene Symbol: ifng1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_212864.1 | NP_998029.1 | 1 - 2 | 120 | 78 | |
| GenBank mRNA | AB158361.1 | - | 1 - 2 | 120 | 78 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ifng1| Gene Symbol: ifng1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_212864.1 | NP_998029.1 | 1 - 2 | 120 | 78 | |
| GenBank mRNA | AB158361.1 | - | 1 - 2 | 120 | 78 | |
Set Membership: |
|