Assay Details
| Gene Symbol: rac3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002754.2 | NP_001002754.1 | 3 - 4 | 344 | 77 | |
| GenBank mRNA | AY865570.1 | - | 3 - 4 | 223 | 77 | |
| BC076433.1 | - | 3 - 4 | 348 | 77 | ||
More Information
Set Membership: |
|

rac3a
Genomic Map