Assay Details
| Gene Symbol: ppp1r7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037386.2 | NP_001032463.1 | 1 - 2 | 58 | 66 | |
| NM_001291498.1 | NP_001278427.1 | 1 - 2 | 58 | 66 | ||
| GenBank mRNA | BC108074.1 | - | 1 - 2 | 60 | 66 | |
| EH446833.1 | - | 1 - 2 | 83 | 66 | ||
| EH537826.1 | - | 1 - 2 | 82 | 66 | ||
More Information
Set Membership: |
|

ppp1r7
Genomic Map