Assay Details
| Gene Symbol: crfb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001079681.1 | NP_001073149.1 | 4 - 5 | 441 | 101 | |
| GenBank mRNA | BC163577.1 | - | 4 - 5 | 414 | 101 | |
| BC163583.1 | - | 4 - 5 | 414 | 101 | ||
| EF014952.1 | - | 4 - 5 | 441 | 101 | ||
More Information
Set Membership: |
|

crfb1
Genomic Map