Assay Details
| Gene Symbol: p2rx1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_198982.1 | NP_945333.1 | 3 - 4 | 365 | 69 | |
| GenBank mRNA | AY292649.1 | - | 3 - 4 | 365 | 69 | |
| BC162912.1 | - | 3 - 4 | 432 | 69 | ||
| BC162955.1 | - | 3 - 4 | 432 | 69 | ||
More Information
Set Membership: |
|

p2rx1
Genomic Map