Assay Details
| Gene Symbol: p2rx3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131623.1 | NP_571698.1 | 1 - 2 | 229 | 111 | |
| GenBank mRNA | AF237683.1 | - | 1 - 2 | 111 | ||
| AF238292.1 | - | 1 - 2 | 229 | 111 | ||
| BC162279.1 | - | 1 - 2 | 215 | 111 | ||
| BC162600.1 | - | 1 - 2 | 215 | 111 | ||
More Information
Set Membership: |
|

p2rx3a
Genomic Map