Assay Details
| Gene Symbol: ndufb8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200855.1 | NP_957149.1 | 3 - 4 | 327 | 67 | |
| GenBank mRNA | BC062842.1 | - | 3 - 4 | 327 | 67 | |
More Information
Set Membership: |
|

ndufb8
Genomic Map