Assay Details
| Gene Symbol: pald1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001089385.1 | NP_001082854.1 | 15 - 16 | 2184 | 73 | |
| XM_005156849.4 | XP_005156906.1 | 15 - 16 | 2187 | 73 | ||
| XM_021480595.1 | XP_021336270.1 | 15 - 16 | 2263 | 73 | ||
| GenBank mRNA | BC139564.1 | - | 15 - 16 | 2184 | 73 | |
More Information
Set Membership: |
|

pald1b
Genomic Map