Assay Details
| Gene Symbol: asic4a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_214787.2 | NP_999952.1 | 4 - 5 | 1235 | 74 | |
| GenBank mRNA | AJ609619.1 | - | 4 - 5 | 1236 | 74 | |
More Information
Set Membership: |
|

asic4a
Genomic Map