Assay Details
| Gene Symbol: asic4a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_214787.2 | NP_999952.1 | 8 - 9 | 1621 | 82 | |
| GenBank mRNA | AJ609619.1 | - | 8 - 9 | 1622 | 82 | |
| Gene Symbol: LOC103910008 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_009298393.2 | XP_009296668.1 | 1 - 2 | 115 | 82 | |
More Information
Set Membership: |
|

asic4a
Genomic Map