Assay Details
| Gene Symbol: rps6kb1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213076.1 | NP_998241.1 | 2 - 3 | 185 | 106 | |
| GenBank mRNA | BC046888.1 | - | 2 - 3 | 185 | 106 | |
| BC165362.1 | - | 2 - 3 | 140 | 106 | ||
More Information
Set Membership: |
|

rps6kb1b
Genomic Map